Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4-Trimethyl-3-pentanol, 98%

$179.31 - $693.11

Chemical Identifiers

Chemical Name or Material 2,3,4-Trimethyl-3-pentanol
CAS 3054-92-0
Molecular Formula C8H18O
Formula Weight 130.23
MDL Number MFCD00048319
Synonym Diisopropyl methyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2023606

  SDS
alfa aesar™
B2023606
5g Each for $179.31

AAB2023614

  SDS
alfa aesar™
B2023614
25g Each for $693.11
Description & Specifications

Specifications

Formula Weight 130.23
Chemical Name or Material 2,3,4-Trimethyl-3-pentanol
Health Hazard 1 Warning
MDL Number MFCD00048319
Quantity 5g
Synonym Diisopropyl methyl carbinol
Molecular Formula C8H18O
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 98%
CAS 3054-92-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.