Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4-Trimethoxybenzonitrile, 97%

$418.85 - $418.85

Chemical Identifiers

Chemical Name or Material 2,3,4-Trimethoxybenzonitrile
CAS 43020-38-8
Molecular Formula C10H11NO3
Formula Weight 193.20g/mol
MDL Number MFCD00001785
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2445414

 
alfa aesar™
B24454-14
25g Each for $418.85
Description & Specifications

Specifications

Melting Point 55° to 57°C
Quantity 25g
Chemical Name or Material 2,3,4-Trimethoxybenzonitrile
CAS 43020-38-8
Assay 97%
Molecular Formula C10H11NO3
Formula Weight 193.20g/mol
MDL Number MFCD00001785
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P280h
Wear protective gloves/clothing.