Learn More
2,3,4-Trimethoxybenzeneboronic acid, 98%
CAS: 118062-05-8 | C9H13BO5 | 212.008 g/mol
$48.03 - $154.47
Chemical Identifiers
| CAS | 118062-05-8 |
|---|---|
| Molecular Formula | C9H13BO5 |
| Molecular Weight (g/mol) | 212.008 |
| MDL Number | MFCD01318183 |
| InChI Key | LDQLSQRFSNMANA-UHFFFAOYSA-N |
| Synonym | 2,3,4-trimethoxyphenyl boronic acid, 2,3,4-trimethoxybenzeneboronic acid, 2,3,4-trimethoxyphenyl boranediol, 2,3,4-trimethoxylphenylboronic acid, boronic acid, 2,3,4-trimethoxyphenyl, pubchem11893, acmc-2099vt, trimethoxyphenyl boronic acid, 2,3,4-trimethoxyphenylboronsaure, ablock ab-12-9176 |
| PubChem CID | 2773582 |
| IUPAC Name | (2,3,4-trimethoxyphenyl)boronic acid |
| SMILES | B(C1=C(C(=C(C=C1)OC)OC)OC)(O)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL1983803
|
Thermo Scientific Chemicals
L1983803 |
1 g |
Each for $48.03
|
|
|||||
|
AAL1983806
|
Thermo Scientific Chemicals
L1983806 |
5 g |
Each for $154.47
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 118062-05-8 | |
| 212.008 | |
| LDQLSQRFSNMANA-UHFFFAOYSA-N | |
| 2773582 | |
| B(C1=C(C(=C(C=C1)OC)OC)OC)(O)O |
| C9H13BO5 | |
| MFCD01318183 | |
| 2,3,4-trimethoxyphenyl boronic acid, 2,3,4-trimethoxybenzeneboronic acid, 2,3,4-trimethoxyphenyl boranediol, 2,3,4-trimethoxylphenylboronic acid, boronic acid, 2,3,4-trimethoxyphenyl, pubchem11893, acmc-2099vt, trimethoxyphenyl boronic acid, 2,3,4-trimethoxyphenylboronsaure, ablock ab-12-9176 | |
| (2,3,4-trimethoxyphenyl)boronic acid |
Specifications
| 118062-05-8 | |
| C9H13BO5 | |
| 1 g | |
| 2,3,4-trimethoxyphenyl boronic acid, 2,3,4-trimethoxybenzeneboronic acid, 2,3,4-trimethoxyphenyl boranediol, 2,3,4-trimethoxylphenylboronic acid, boronic acid, 2,3,4-trimethoxyphenyl, pubchem11893, acmc-2099vt, trimethoxyphenyl boronic acid, 2,3,4-trimethoxyphenylboronsaure, ablock ab-12-9176 | |
| B(C1=C(C(=C(C=C1)OC)OC)OC)(O)O | |
| 212.008 | |
| 212.01 | |
| 2,3,4-Trimethoxybenzeneboronic acid |
| 74°C to 79°C | |
| MFCD01318183 | |
| 4185924 | |
| LDQLSQRFSNMANA-UHFFFAOYSA-N | |
| (2,3,4-trimethoxyphenyl)boronic acid | |
| 2773582 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only