Please login to your online account to display your discounted pricing

2,3,4-Trimethoxybenzaldehyde, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3, 4-Trimethoxybenzaldehyde
Name Note 99%
CAS 2103-57-3
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C10H12O4
Linear Formula (CH3O)3C6H2CHO
Formula Weight 196.2
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC166450100

  SDS
fisher scientific acros
166450100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC166450500

  SDS
fisher scientific acros
166450500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.2
Melting Point >36.5°C
Assay ≥98.5%
Physical Form Crystals or Crystalline Powder
Quantity 10g
Color White to light yellow
Beilstein 08, I, 684
TSCA TSCA
EINECS Number 218-271-0
CAS Max % 100.0
Infrared Spectrum Authentic
CAS Min % 98.5
Packaging Glass bottle
Chemical Name or Material 2, 3, 4-Trimethoxybenzaldehyde
Molecular Formula C10H12O4
Name Note 99%
Linear Formula (CH3O)3C6H2CHO
Assay Percent Range 99%
MDL Number MFCD00003310
CAS 2103-57-3