Please login to your online account to display your discounted pricing

2',3',4'-Trimethoxyacetophenone, 96%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  183490050

 View more versions of this product

Catalog No. AC183490050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5374 to 1.5394 (20°C, 589nm)
Density 1.1500g/mL
Color Clear yellow
Quantity 5mL
Chemical Name or Material 2', 3', 4'-Trimethoxyacetophenone
Name Note 96%
CAS 13909-73-4
CAS Min % 95.0
CAS Max % 100.0
Physical Form Clear yellow liquid
Assay 95% min.
Assay Percent Range 96%
Molecular Formula C11 H14 O4
Linear Formula (CH3O)3C6H2COCH3
Formula Weight 210.23g/mol
MDL Number MFCD00008726
Beilstein 08, 393
EINECS Number 237-678-4
TSCA TSCA

Specifications

Chemical Name or Material 2', 3', 4'-Trimethoxyacetophenone
Name Note 96%
CAS 13909-73-4
CAS Min % 95.0
CAS Max % 100.0
Assay 95% min.
Assay Percent Range 96%
Molecular Formula C11 H14 O4
Linear Formula (CH3O)3C6H2COCH3
Formula Weight 210.23g/mol
MDL Number MFCD00008726