Please login to your online account to display your discounted pricing

2',3',4'-Trimethoxyacetophenone, 96%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2', 3', 4'-Trimethoxyacetophenone
Name Note 96%
CAS 13909-73-4
CAS Min % 95.0
CAS Max % 100.0
Assay 95% min.
Assay Percent Range 96%
Molecular Formula C11 H14 O4
Linear Formula (CH3O)3C6H2COCH3
Formula Weight 210.23g/mol
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC183490050

  SDS
thermo fisher scientific
183490050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5374 to 1.5394 (20°C, 589nm)
Density 1.1500g/mL
Color Clear yellow
Quantity 5mL
Chemical Name or Material 2', 3', 4'-Trimethoxyacetophenone
Name Note 96%
CAS 13909-73-4
CAS Min % 95.0
CAS Max % 100.0
Physical Form Clear yellow liquid
Assay 95% min.
Assay Percent Range 96%
Molecular Formula C11 H14 O4
Linear Formula (CH3O)3C6H2COCH3
Formula Weight 210.23g/mol
MDL Number MFCD00008726
Beilstein 08, 393
EINECS Number 237-678-4
TSCA TSCA