Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4-Trihydroxybenzophenone, 98%

$66.91 - $846.80

Chemical Identifiers

Chemical Name or Material 2,3,4-Trihydroxybenzophenone
CAS 1143-72-2
Molecular Formula C13H10O4
Formula Weight 230.22
MDL Number MFCD00009996
Beilstein 2697065
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1831514

  SDS
alfa aesar™
A1831514
25g Each for $66.91

AAA1831522

  SDS
alfa aesar™
A1831522
100g Each for $180.81

AAA1831536

  SDS
alfa aesar™
A1831536
500g Each for $846.80
Description & Specifications

Specifications

Formula Weight 230.22
Chemical Name or Material 2,3,4-Trihydroxybenzophenone
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
CAS 1143-72-2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 140° to 142°C
MDL Number MFCD00009996
Molecular Formula C13H10O4
Health Hazard 1 Warning
Assay 98%
Beilstein 2697065
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.