Learn More
2,3,4-Trihydroxybenzoic acid, 97%, Thermo Scientific Chemicals
$51.37 - $555.55
Chemical Identifiers
CAS | 610-02-6 |
---|---|
Molecular Formula | C7H6O5 |
Molecular Weight (g/mol) | 170.12 |
MDL Number | MFCD00002447 |
InChI Key | BRRSNXCXLSVPFC-UHFFFAOYSA-N |
Synonym | 4-pyrogallolcarboxylic acid, pyrogallolcarboxylic acid, 2,3,4-trihydroxybenzoate, unii-ad1id2jf5o, benzoic acid, trihydroxy, benzoic acid, 2,3,4-trihydroxy, pyrogallol-4-carboxylic acid, 2,3,4-trihydroxybenzene carboxylic acid, ad1id2jf5o, pyrogallolcarboxylate |
PubChem CID | 11874 |
IUPAC Name | 2,3,4-trihydroxybenzoic acid |
SMILES | OC(=O)C1=CC=C(O)C(O)=C1O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2037103
|
Thermo Scientific Chemicals
B2037103 |
1 g |
Each for $51.37
|
|
|||||
AAB2037106
|
Thermo Scientific Chemicals
B2037106 |
5 g |
Each for $178.96
|
|
|||||
AAB2037114
|
Thermo Scientific Chemicals
B2037114 |
25 g |
Each for $555.55
|
|
|||||
Description
2,3,4-Trihydroxybenzoic acid is used as a reagent during catalytic spectrophotometric determination of vanadium in natural and sea waters and used in the pharmaceuticals. It is a internal standard in separation of phenolic acids by HPLC with photodiode array detection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,3,4-Trihydroxybenzoic acid is used as a reagent during catalytic spectrophotometric determination of vanadium in natural and sea waters and used in the pharmaceuticals. It is a internal standard in separation of phenolic acids by HPLC with photodiode array detection.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
Chemical Identifiers
610-02-6 | |
170.12 | |
BRRSNXCXLSVPFC-UHFFFAOYSA-N | |
11874 | |
OC(=O)C1=CC=C(O)C(O)=C1O |
C7H6O5 | |
MFCD00002447 | |
4-pyrogallolcarboxylic acid, pyrogallolcarboxylic acid, 2,3,4-trihydroxybenzoate, unii-ad1id2jf5o, benzoic acid, trihydroxy, benzoic acid, 2,3,4-trihydroxy, pyrogallol-4-carboxylic acid, 2,3,4-trihydroxybenzene carboxylic acid, ad1id2jf5o, pyrogallolcarboxylate | |
2,3,4-trihydroxybenzoic acid |
Specifications
610-02-6 | |
C7H6O5 | |
1 g | |
4-pyrogallolcarboxylic acid, pyrogallolcarboxylic acid, 2,3,4-trihydroxybenzoate, unii-ad1id2jf5o, benzoic acid, trihydroxy, benzoic acid, 2,3,4-trihydroxy, pyrogallol-4-carboxylic acid, 2,3,4-trihydroxybenzene carboxylic acid, ad1id2jf5o, pyrogallolcarboxylate | |
BRRSNXCXLSVPFC-UHFFFAOYSA-N | |
2,3,4-trihydroxybenzoic acid | |
11874 | |
97% |
∼210°C (decomposition) | |
MFCD00002447 | |
2210807 | |
Slightly soluble in water. | |
OC(=O)C1=CC=C(O)C(O)=C1O | |
170.12 | |
170.12 | |
2,3,4-Trihydroxybenzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 210-205-9
RTECSNumber : DH8900000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only