missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,4-Trihydroxybenzaldehyde, TCI America™

Manufacturer:  TCI America T21585G

 View more versions of this product

Catalog No. T21585G



Specifications

Specifications

2,3,4-Trihydroxybenzaldehyde
162°C
2144-08-3
pyrogallol-4-carboxaldehyde, benzaldehyde, 2,3,4-trihydroxy, pyrogallolaldehyd, pubchem8260, 2,4-trihydroxybenzaldehyde, dsstox_cid_31473, dsstox_rid_97358, dsstox_gsid_57684, ksc490m2j, benzaldehyde,3,4-trihydroxy
C1=CC(=C(C(=C1C=O)O)O)O
154.12
154.12
≥98.0% (GC,T)
Undesignated
5g
C7H6O4
CRPNQSVBEWWHIJ-UHFFFAOYSA-N
2,3,4-trihydroxybenzaldehyde
75064
Crystal-Powder at 20°C

Chemical Identifiers

2144-08-3
154.12
pyrogallol-4-carboxaldehyde, benzaldehyde, 2,3,4-trihydroxy, pyrogallolaldehyd, pubchem8260, 2,4-trihydroxybenzaldehyde, dsstox_cid_31473, dsstox_rid_97358, dsstox_gsid_57684, ksc490m2j, benzaldehyde,3,4-trihydroxy
2,3,4-trihydroxybenzaldehyde
C7H6O4
CRPNQSVBEWWHIJ-UHFFFAOYSA-N
75064
C1=CC(=C(C(=C1C=O)O)O)O
Safety and Handling

Safety and Handling

RTECSNumber : CU8439800

TSCA : Yes

Documents