missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',3',4'-Trihydroxyacetophenone, 98%

$61.19 - $592.38

Chemical Identifiers

CAS 528-21-2
Molecular Formula C8H8O4
Formula Weight 168.15
MDL Number MFCD00002193
Merck Index 14,4342
Synonym 4-Acetylpyrogallol
Chemical Name or Material 2',3',4'-Trihydroxyacetophenone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1436406 Alfa Aesar™
A1436406
5g Each for $61.19
AAA1436414 Alfa Aesar™
A1436414
25g Each for $169.04
AAA1436422 Alfa Aesar™
A1436422
100g Each for $592.38
Specifications Description & Specifications

Chemical Identifiers

528-21-2
168.15
14,4342
2',3',4'-Trihydroxyacetophenone
C8H8O4
MFCD00002193
4-Acetylpyrogallol

Specifications

169° to 172°C
528-21-2
168.15
14,4342
2',3',4'-Trihydroxyacetophenone
5g
C8H8O4
MFCD00002193
4-Acetylpyrogallol
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : AN0527000

TSCA : TSCA