missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,4-Trifluorotoluene, TCI America™

Manufacturer:  TCI America T27351G

 View more versions of this product

Catalog No. T27351G



Specifications

Specifications

2,3,4-Trifluorotoluene
Yellow
193533-92-5
MFCD00075194
2,3,4-trifluorotoluene, benzene, 1,2,3-trifluoro-4-methyl, triflurotoluene, 1,2,3-trifluoro-4-methyl-benzene, pubchem1628, acmc-1cn3t, ksc541e8d, 2,3,4-trifluorotoluene, benzene,1,2,3-trifluoro-4-methyl, benzene, 1,2,3-trifluoro-4-methyl-9ci
CC1=C(C(=C(C=C1)F)F)F
146.112
146.11
≥97.0% (GC)
127°C
1g
C7H5F3
1993
LRQPEHJWTXCLQY-UHFFFAOYSA-N
1,2,3-trifluoro-4-methylbenzene
2777991
Clear Liquid at 20°C

Chemical Identifiers

193533-92-5
146.112
LRQPEHJWTXCLQY-UHFFFAOYSA-N
2777991
CC1=C(C(=C(C=C1)F)F)F
C7H5F3
MFCD00075194
2,3,4-trifluorotoluene, benzene, 1,2,3-trifluoro-4-methyl, triflurotoluene, 1,2,3-trifluoro-4-methyl-benzene, pubchem1628, acmc-1cn3t, ksc541e8d, 2,3,4-trifluorotoluene, benzene,1,2,3-trifluoro-4-methyl, benzene, 1,2,3-trifluoro-4-methyl-9ci
1,2,3-trifluoro-4-methylbenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents