Please login to your online account to display your discounted pricing

2,3,4-Trifluorophenol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3, 4-Trifluorophenol
CAS 2822-41-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H3F3O
Linear Formula F3C6H2OH
Formula Weight 148.09
MDL Number MFCD00042426
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC308550010

  SDS
thermo fisher scientific
308550010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid After Melting
Formula Weight 148.09
Quantity 1g
Color Clear colorless to slightly yellow
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2, 3, 4-Trifluorophenol
Assay Percent Range 97%
Molecular Formula C6H3F3O
CAS 2822-41-5
Infrared Spectrum Authentic
Packaging Glass bottle
MDL Number MFCD00042426
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4635 to 1.4655 (20°C, 589nm)
Linear Formula F3C6H2OH
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable solid.
Flammable liquid and vapor.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Keep container tightly closed.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Flammable solid.
Flammable liquid and vapor.