missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,4-Trifluorobenzyl bromide, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC442260050

 View more versions of this product

Catalog No. AC442260050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 3, 4-Trifluorobenzyl bromide
Authentic
5g
96.0
96.5% min. (GC)
1-bromomethyl-2,3,4-trifluorobenzene, 2, 3, 4-trifluorobenzyl bromide, 2,3,4-trifluoro benzyl bromide, 2,3,4-trifluorobenzyl bromide, 2,3,4-trifluorobenzylbromide, 2,3,4-trifluorophenyl methyl bromide, 4-bromomethyl-1,2,3-trifluorobenzene, 6-bromomethyl-1,2,3-trifluorobenzene, alpha-bromo-2,3,4-trifluorotoluene, benzene, 1-bromomethyl-2,3,4-trifluoro
C1=CC(=C(C(=C1CBr)F)F)F
225.01
225.01
97%
90°C
Colorless to Yellow
157911-55-2
100.0
C7H4BrF3
DGSXDQVPGXFOAN-UHFFFAOYSA-N
1-(bromomethyl)-2,3,4-trifluorobenzene
2777050
Liquid

Chemical Identifiers

157911-55-2
225.01
1-bromomethyl-2,3,4-trifluorobenzene, 2, 3, 4-trifluorobenzyl bromide, 2,3,4-trifluoro benzyl bromide, 2,3,4-trifluorobenzyl bromide, 2,3,4-trifluorobenzylbromide, 2,3,4-trifluorophenyl methyl bromide, 4-bromomethyl-1,2,3-trifluorobenzene, 6-bromomethyl-1,2,3-trifluorobenzene, alpha-bromo-2,3,4-trifluorotoluene, benzene, 1-bromomethyl-2,3,4-trifluoro
1-(bromomethyl)-2,3,4-trifluorobenzene
C7H4BrF3
DGSXDQVPGXFOAN-UHFFFAOYSA-N
2777050
C1=CC(=C(C(=C1CBr)F)F)F
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents