Please login to your online account to display your discounted pricing

2,3,4-Trifluorobenzyl bromide, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3, 4-Trifluorobenzyl bromide
CAS 157911-55-2
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.5%
Assay Percent Range 97%
Molecular Formula C7H4BrF3
Formula Weight 225.01
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC442260010

  SDS
thermo fisher scientific
AC442260010
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC442260050

  SDS
thermo fisher scientific
AC442260050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.5%
Formula Weight 225.01
Physical Form Liquid
Quantity 1g
Color Clear colorless to pale yellow
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C7H4BrF3
Assay Percent Range 97%
CAS 157911-55-2
Chemical Name or Material 2, 3, 4-Trifluorobenzyl bromide
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.