Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4-Trifluorobenzyl alcohol, 97%

$76.39 - $263.69

Chemical Identifiers

Chemical Name or Material 2,3,4-Trifluorobenzyl alcohol
CAS 144284-24-2
Molecular Formula C7H5F3O
Formula Weight 162.11
MDL Number MFCD00083551
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2430603

  SDS
alfa aesar™
B2430603
1g Each for $76.39

AAB2430606

  SDS
alfa aesar™
B2430606
5g Each for $263.69
Description & Specifications

Specifications

Formula Weight 162.11
Molecular Formula C7H5F3O
Health Hazard 1 Warning
MDL Number MFCD00083551
CAS 144284-24-2
Assay 97%
Quantity 1g
Chemical Name or Material 2,3,4-Trifluorobenzyl alcohol
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.