Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4-Trifluorobenzonitrile, 98%

Chemical Identifiers

Chemical Name or Material 2,3,4-Trifluorobenzonitrile
CAS 143879-80-5
Molecular Formula C7H2F3N
Formula Weight 157.1
MDL Number MFCD00013288
UN Number UN3276
Beilstein 5427682
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2069103

  SDS
alfa aesar™
B2069103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2069106

  SDS
alfa aesar™
B2069106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 157.1
Flash Point 68°C (154°F)
Quantity 1g
Chemical Name or Material 2,3,4-Trifluorobenzonitrile
MDL Number MFCD00013288
Molecular Formula C7H2F3N
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.
Health Hazard 2 GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 116° to 118°C (100mmHg)
UN Number UN3276
CAS 143879-80-5
Health Hazard 1 Danger
Assay 98%
Beilstein 5427682
Safety and Handling

Danger

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.