Please login to your online account to display your discounted pricing

2,3,4-Trifluorobenzene-1-sulfonyl chloride, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2, 3, 4-Trifluorobenzene-1-sulfonyl chloride
CAS 175278-08-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H2ClF3O2S
Linear Formula F3C6H2SO2Cl
Formula Weight 230.59
MDL Number MFCD00102573
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

TL00605EA

 
fisher scientific company
TL00605EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 230.59
Quantity 10g
Assay 97%
Density 1.6400g/mL
CAS Min % 97.0
Chemical Name or Material 2, 3, 4-Trifluorobenzene-1-sulfonyl chloride
Packaging Amber glass bottle
Molecular Formula C6H2ClF3O2S
Assay Percent Range 97%
MDL Number MFCD00102573
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula F3C6H2SO2Cl
CAS 175278-08-7
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.