Please login to your online account to display your discounted pricing

2,3,4-Trifluorobenzene-1-sulfonyl chloride, 97%, Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  TL00605EA

 View more versions of this product

Catalog No. TL00605EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 230.59
Quantity 10g
Assay 97%
Density 1.6400g/mL
CAS Min % 97.0
Chemical Name or Material 2, 3, 4-Trifluorobenzene-1-sulfonyl chloride
Packaging Amber glass bottle
Molecular Formula C6H2ClF3O2S
Assay Percent Range 97%
MDL Number MFCD00102573
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula F3C6H2SO2Cl
CAS 175278-08-7
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

Specifications

Chemical Name or Material 2, 3, 4-Trifluorobenzene-1-sulfonyl chloride
CAS 175278-08-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H2ClF3O2S
Linear Formula F3C6H2SO2Cl
Formula Weight 230.59
MDL Number MFCD00102573
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.