missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,4-Trifluorobenzaldehyde, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC312710050

 View more versions of this product

Catalog No. AC312710050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
160.09
Clear colorless
100.0
C7H3F3O
2, 3, 4-Trifluorobenzaldehyde
Authentic
1.4800 to 1.4820 (20°C, 589nm)
98%
Liquid
5g
1.4040g/mL
F3C6H2CHO
MFCD00061230
97.5
Glass bottle
161793-17-5

Chemical Identifiers

161793-17-5
100.0
C7H3F3O
160.09
2, 3, 4-Trifluorobenzaldehyde
97.5
98%
F3C6H2CHO
MFCD00061230
≥97.5%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning