Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4-Trifluorobenzaldehyde, 98%

$88.05 - $88.05

Chemical Identifiers

Chemical Name or Material 2,3,4-Trifluorobenzaldehyde
CAS 161793-17-5
Molecular Formula C7H3F3O
Formula Weight 160.1
MDL Number MFCD00061230
Beilstein 7805818
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1995503

  SDS
alfa aesar™
A1995503
1g Each for $88.05
Description & Specifications

Specifications

Formula Weight 160.1
Chemical Name or Material 2,3,4-Trifluorobenzaldehyde
Assay 98%
Molecular Formula C7H3F3O
MDL Number MFCD00061230
CAS 161793-17-5
Beilstein 7805818
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Flash Point 61°C (142°F)
Boiling Point 77° to 79°C (20mmHg)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.