Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4-Trifluoroanisole, 97+%

$60.66 - $213.96

Chemical Identifiers

Chemical Name or Material 2,3,4-Trifluoroanisole
CAS 203245-16-3
Molecular Formula C7H5F3O
Formula Weight 162.11
MDL Number MFCD00042396
Synonym 1-Methoxy-2,3,4-trifluorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2401403

 
alfa aesar™
B2401403
1g Each for $60.66

AAB2401406

 
alfa aesar™
B2401406
5g Each for $213.96
Description & Specifications

Specifications

Formula Weight 162.11
Chemical Name or Material 2,3,4-Trifluoroanisole
Assay ≥97%
Molecular Formula C7H5F3O
MDL Number MFCD00042396
CAS 203245-16-3
Synonym 1-Methoxy-2,3,4-trifluorobenzene
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H227
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 61°C (142°F)
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H227
Combustible liquid.