missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,4-Trifluoroaniline, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 268930050

 View more versions of this product

Catalog No. AC268930050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2, 3, 4-Trifluoroaniline
68°C
Glass bottle
1.39
Brown to Purple
98%
97.5
98%
F3C6H2NH2
12, 114
WRDGNXCXTDDYBZ-UHFFFAOYSA-N
2,3,4-trifluoroaniline
77468
Liquid
1.3900g/mL
Authentic
1.4860 to 1.488
92°C (48.0mmHg)
5g
3862-73-5
100.0
C6H4F3N
MFCD00011737
2,3,4-trifluoraniline, 2,3,4-trifluoro aniline, 2,3,4-trifluoro-phenylamine, 2,3,4-trifluorobenzenamine, 2,3,4-trifluorophenylamine, 2.3.4-trifluoroaniline, acmc-1clgf, aniline, 2,3,4-trifluoro, benzenamine, 2,3,4-trifluoro, pubchem2297
C1=CC(=C(C(=C1N)F)F)F
147.1
147.1
98%

Chemical Identifiers

3862-73-5
147.1
WRDGNXCXTDDYBZ-UHFFFAOYSA-N
77468
C1=CC(=C(C(=C1N)F)F)F
C6H4F3N
MFCD00011737
2,3,4-trifluoraniline, 2,3,4-trifluoro aniline, 2,3,4-trifluoro-phenylamine, 2,3,4-trifluorobenzenamine, 2,3,4-trifluorophenylamine, 2.3.4-trifluoroaniline, acmc-1clgf, aniline, 2,3,4-trifluoro, benzenamine, 2,3,4-trifluoro, pubchem2297
2,3,4-trifluoroaniline
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in c

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:

GHS Signal Word: Danger

RTECSNumber : CY1211500

TSCA : TSCA

SDS Documents