Please login to your online account to display your discounted pricing

2,3,4-Trifluoroaniline, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2, 3, 4-Trifluoroaniline
Name Note 98%
CAS 3862-73-5
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H4F3N
Linear Formula F3C6H2NH2
Formula Weight 147.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC268930050

  SDS
thermo fisher scientific
268930050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 147.1
Physical Form Liquid
Quantity 5g
Color Clear purple to brown
TSCA TSCA
RTECS Number CY1211500
Density 1.3900g/mL
Beilstein 12, 114
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 2 GHS P Statement
Toxic to aquatic life with long lasting effects.
Causes serious eye damage.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 1 Danger
CAS Min % 97.5
Packaging Glass bottle
Synonym 2, 3, 4-Trifluorobenzenamine
Refractive Index 1.4860 to 1.4880 (20°C, 589nm)
Health Hazard 3 GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula F3C6H2NH2
MDL Number MFCD00011737
Assay Percent Range 98%
Molecular Formula C6H4F3N
Chemical Name or Material 2, 3, 4-Trifluoroaniline
Name Note 98%
CAS 3862-73-5