missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4-Trifluoroaniline, 98%

$151.76 - $522.78

Chemical Identifiers

CAS 3862-73-5
Molecular Formula C6H4F3N
Molecular Weight (g/mol) 147.1
MDL Number MFCD00011737
InChI Key WRDGNXCXTDDYBZ-UHFFFAOYSA-N
Synonym 2,3,4-trifluorobenzenamine, benzenamine, 2,3,4-trifluoro, 2,3,4-trifluoraniline, 2,3,4-trifluoro-phenylamine, aniline, 2,3,4-trifluoro, 2,3,4-trifluoro aniline, 2,3,4-trifluorophenylamine, pubchem2297, acmc-1clgf, 2.3.4-trifluoroaniline
PubChem CID 77468
IUPAC Name 2,3,4-trifluoroaniline
SMILES C1=CC(=C(C(=C1N)F)F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1025422 View Documents Alfa Aesar™
A1025422
100g Each for $522.78
 
AAA1025414 View Documents Alfa Aesar™
A1025414
25g Each for $151.76
 
Specifications

Chemical Identifiers

3862-73-5
147.1
WRDGNXCXTDDYBZ-UHFFFAOYSA-N
77468
C1=CC(=C(C(=C1N)F)F)F
C6H4F3N
MFCD00011737
2,3,4-trifluorobenzenamine, benzenamine, 2,3,4-trifluoro, 2,3,4-trifluoraniline, 2,3,4-trifluoro-phenylamine, aniline, 2,3,4-trifluoro, 2,3,4-trifluoro aniline, 2,3,4-trifluorophenylamine, pubchem2297, acmc-1clgf, 2.3.4-trifluoroaniline
2,3,4-trifluoroaniline

Specifications

68°C (154°F)
100g
C6H4F3N
UN3082
2,3,4-trifluorobenzenamine, benzenamine, 2,3,4-trifluoro, 2,3,4-trifluoraniline, 2,3,4-trifluoro-phenylamine, aniline, 2,3,4-trifluoro, 2,3,4-trifluoro aniline, 2,3,4-trifluorophenylamine, pubchem2297, acmc-1clgf, 2.3.4-trifluoroaniline
C1=CC(=C(C(=C1N)F)F)F
147.1
147.1
2,3,4-Trifluoroaniline
90°C to 92°C (48mmHg)
3862-73-5
MFCD00011737
3245609
WRDGNXCXTDDYBZ-UHFFFAOYSA-N
2,3,4-trifluoroaniline
77468
98%
Safety and Handling

Safety and Handling

GHS H Statement
H318-H373-H302-H312-H227-H315
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Combustible liquid.
Causes skin irritation.

GHS P Statement
P210-P260-P280-P305+P351+P338-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.

RTECSNumber : CY1211500