Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4-Trifluoro-6-nitroaniline, 98%

Chemical Identifiers

Chemical Name or Material 2,3,4-Trifluoro-6-nitroaniline
CAS 148416-38-0
Formula Weight 192.1
MDL Number MFCD00174089
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6151406

 
alfa aesar™
H6151406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6151403

 
alfa aesar™
H6151403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 59° to 61°C
Formula Weight 192.1
Assay 98%
Quantity 5g
MDL Number MFCD00174089
Physical Form Powder
CAS 148416-38-0
Color White
Chemical Name or Material 2,3,4-Trifluoro-6-nitroaniline