missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4-Trichlorophenyl isothiocyanate, 97%

Manufacturer:  Alfa Aesar™ L1211906

 View more versions of this product

Catalog No. AAL1211906



Specifications

Specifications

2,3,4-Trichlorophenyl isothiocyanate
5g
C7H2Cl3NS
UN2923
2,3,4-trichlorophenyl isothiocyanate, benzene,1,2,3-trichloro-4-isothiocyanato, acmc-1bzch, trichloro phenyl isothiocyanate, 2,3,4-trichlorobenzenisothiocyanate, 2,3,4-trichlorophenylisothiocyanate, #, benzene, 1,2,3-trichloro-4-isothiocyanato-9ci
C1=CC(=C(C(=C1N=C=S)Cl)Cl)Cl
238.51
238.52
68°C to 71°C
127142-69-2
MFCD00041049
6921596
NUAGIDXAYVLNEG-UHFFFAOYSA-N
1,2,3-trichloro-4-isothiocyanatobenzene
145582
97%

Chemical Identifiers

127142-69-2
238.51
NUAGIDXAYVLNEG-UHFFFAOYSA-N
145582
C1=CC(=C(C(=C1N=C=S)Cl)Cl)Cl
C7H2Cl3NS
MFCD00041049
2,3,4-trichlorophenyl isothiocyanate, benzene,1,2,3-trichloro-4-isothiocyanato, acmc-1bzch, trichloro phenyl isothiocyanate, 2,3,4-trichlorobenzenisothiocyanate, 2,3,4-trichlorophenylisothiocyanate, #, benzene, 1,2,3-trichloro-4-isothiocyanato-9ci
1,2,3-trichloro-4-isothiocyanatobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

Documents