Please login to your online account to display your discounted pricing

Alfa Aesar™ 2',3',4'-Trichloroacetophenone, 95%

$35.85 - $289.30

Chemical Identifiers

Chemical Name or Material 2',3',4'-Trichloroacetophenone
CAS 13608-87-2
Molecular Formula C8H5Cl3O
Formula Weight 223.49
MDL Number MFCD00000542
Beilstein 1948674
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1379006

  SDS
alfa aesar™
A1379006
5g Each for $35.85

AAA1379014

  SDS
alfa aesar™
A1379014
25g Each for $88.15

AAA1379022

  SDS
alfa aesar™
A1379022
100g Each for $289.30
Description & Specifications

Specifications

Formula Weight 223.49
CAS 13608-87-2
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 2',3',4'-Trichloroacetophenone
Quantity 5g
Molecular Formula C8H5Cl3O
Beilstein 1948674
Health Hazard 1 Warning
Assay 95%
MDL Number MFCD00000542
Melting Point 60° to 64°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.