missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3',4'-Trichloroacetophenone

Chemical Identifiers

CAS 42981-08-8
Molecular Formula C8H5Cl3O
Formula Weight 223.49
MDL Number MFCD03970382
UN Number UN1759
Synonym 3,4-Dichlorophenacyl chloride
Chemical Name or Material 2,3',4'-Trichloroacetophenone
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2539706 SDS Alfa Aesar™
B2539706
5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAB2539714 SDS Alfa Aesar™
B2539714
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Applications
2,3′,4′-Trichloroacetophenone is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
Description & Specifications

Chemical Identifiers

42981-08-8
223.49
UN1759
2,3',4'-Trichloroacetophenone
C8H5Cl3O
MFCD03970382
3,4-Dichlorophenacyl chloride

Specifications

42981-08-8
223.49
UN1759
43° to 45°C
5g
C8H5Cl3O
MFCD03970382
3,4-Dichlorophenacyl chloride
171° to 173°C (15mmHg)
2,3',4'-Trichloroacetophenone

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.