Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4-Tribromothiophene, 97%

Chemical Identifiers

Chemical Name or Material 2,3,4-Tribromothiophene
CAS 3141-25-1
Molecular Formula C4HBr3S
Formula Weight 320.84
MDL Number MFCD00041210
UN Number UN2811
Beilstein 111770
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0394603

 
alfa aesar™
L0394603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0394606

 
alfa aesar™
L0394606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 320.84
Chemical Name or Material 2,3,4-Tribromothiophene
Assay 97%
Molecular Formula C4HBr3S
MDL Number MFCD00041210
CAS 3141-25-1
Beilstein 111770
Melting Point 43° to 45°C
Boiling Point 150° to 152°C (20mmHg)
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN2811
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.