Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4-Tri-O-benzyl-beta-L-arabinopyranose, 97%

$107.30 - $304.78

Chemical Identifiers

Chemical Name or Material 2,3,4-Tri-O-benzyl-beta-L-arabinopyranose
CAS 77943-33-0
Molecular Formula C26H28O5
Formula Weight 420.51
MDL Number MFCD05664044
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1979003

 
alfa aesar™
L1979003
1g Each for $304.78

AAL19790MD

 
alfa aesar™
L19790MD
250mg Each for $107.30
Description & Specifications

Specifications

Formula Weight 420.51
Assay 97%
Molecular Formula C26H28O5
Recommended Storage 2° to 8°C
Melting Point 76° to 88°C
Chemical Name or Material 2,3,4-Tri-O-benzyl-beta-L-arabinopyranose
MDL Number MFCD05664044
CAS 77943-33-0
Optical Rotation +78° (c=1 in Methanol)
Quantity 1g

Applications
2,3,4-Tri-O-benzyl-beta-L-arabinopyranose is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Safety and Handling

2° to 8°C