Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4-Tri-O-benzyl-beta-D-arabinopyranose, 97+%

$367.65 - $367.65

Chemical Identifiers

Chemical Name or Material 2,3,4-Tri-O-benzyl-beta-D-arabinopyranose
CAS 18039-26-4
Molecular Formula C26H28O5
Formula Weight 420.51
MDL Number MFCD05664045
Beilstein 5303314
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1987003

  SDS
alfa aesar™
L1987003
1g Each for $367.65
Description & Specifications

Specifications

Formula Weight 420.51
Chemical Name or Material 2,3,4-Tri-O-benzyl-beta-D-arabinopyranose
MDL Number MFCD05664045
Recommended Storage 2° to 8°C
Optical Rotation −70° (c=1 in Methanol)
Molecular Formula C26H28O5
Assay ≥97%
Quantity 1g
Beilstein 5303314
Melting Point 73° to 75°C
CAS 18039-26-4
Safety and Handling

2° to 8°C