missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[3,4-(Methylenedioxy)phenyl]ethylamine hydrochloride, 95%, Alfa Aesar™

$61.38 - $246.74

Chemical Identifiers

CAS 1653-64-1
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD00035193
InChI Key NDYXFQODWGEGNU-UHFFFAOYSA-N
Synonym 3,4-methylenedioxyphenethylamine hydrochloride, 2-3,4-methylenedioxyphenyl ethylamine hydrochloride, phenethylamine, 3,4-methylenedioxy-, hydrochloride, 2-benzo d 1,3 dioxol-5-yl ethanamine hydrochloride, homopiperonylamine hydrochloride, 1,3-benzodioxole-5-ethanamine, hydrochloride, 3,4-methylenedioxy-beta-phenylethylamine hydrochloride, methylenedioxyphenethylamine hydrochloride, 3,4-methylenedioxy phenylethylamine hydrochloride, 3,4-methylenedioxy-.beta.-phenylethylamine hydrochloride
PubChem CID 74246
IUPAC Name 2-(1,3-benzodioxol-5-yl)ethanamine;hydrochloride
SMILES C1OC2=C(O1)C=C(C=C2)CCN.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6020803 View Documents Alfa Aesar
H6020803
1g Each for $61.38
 
AAH6020806 View Documents Alfa Aesar
H6020806
5g Each for $246.74
 
Specifications

Chemical Identifiers

1653-64-1
201.65
NDYXFQODWGEGNU-UHFFFAOYSA-N
74246
C1OC2=C(O1)C=C(C=C2)CCN.Cl
C9H12ClNO2
MFCD00035193
3,4-methylenedioxyphenethylamine hydrochloride, 2-3,4-methylenedioxyphenyl ethylamine hydrochloride, phenethylamine, 3,4-methylenedioxy-, hydrochloride, 2-benzo d 1,3 dioxol-5-yl ethanamine hydrochloride, homopiperonylamine hydrochloride, 1,3-benzodioxole-5-ethanamine, hydrochloride, 3,4-methylenedioxy-beta-phenylethylamine hydrochloride, methylenedioxyphenethylamine hydrochloride, 3,4-methylenedioxy phenylethylamine hydrochloride, 3,4-methylenedioxy-.beta.-phenylethylamine hydrochloride
2-(1,3-benzodioxol-5-yl)ethanamine;hydrochloride

Specifications

213°C to 214°C
1653-64-1
MFCD00035193
NDYXFQODWGEGNU-UHFFFAOYSA-N
2-(1,3-benzodioxol-5-yl)ethanamine;hydrochloride
74246
95%
1g
C9H12ClNO2
3,4-methylenedioxyphenethylamine hydrochloride, 2-3,4-methylenedioxyphenyl ethylamine hydrochloride, phenethylamine, 3,4-methylenedioxy-, hydrochloride, 2-benzo d 1,3 dioxol-5-yl ethanamine hydrochloride, homopiperonylamine hydrochloride, 1,3-benzodioxole-5-ethanamine, hydrochloride, 3,4-methylenedioxy-beta-phenylethylamine hydrochloride, methylenedioxyphenethylamine hydrochloride, 3,4-methylenedioxy phenylethylamine hydrochloride, 3,4-methylenedioxy-.beta.-phenylethylamine hydrochloride
C1OC2=C(O1)C=C(C=C2)CCN.Cl
201.65
201.65
2-[3,4-(Methylenedioxy)phenyl]ethylamine hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : SH9710000