missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-[3,4-(Methylenedioxy)phenyl]ethylamine hydrochloride, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6020806
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-[3,4-(Methylenedioxy)phenyl]ethylamine hydrochloride | |
213°C to 214°C | |
MFCD00035193 | |
3,4-methylenedioxyphenethylamine hydrochloride, 2-3,4-methylenedioxyphenyl ethylamine hydrochloride, phenethylamine, 3,4-methylenedioxy-, hydrochloride, 2-benzo d 1,3 dioxol-5-yl ethanamine hydrochloride, homopiperonylamine hydrochloride, 1,3-benzodioxole-5-ethanamine, hydrochloride, 3,4-methylenedioxy-beta-phenylethylamine hydrochloride, methylenedioxyphenethylamine hydrochloride, 3,4-methylenedioxy phenylethylamine hydrochloride, 3,4-methylenedioxy-.beta.-phenylethylamine hydrochloride | |
C1OC2=C(O1)C=C(C=C2)CCN.Cl | |
201.65 | |
201.65 |
1653-64-1 | |
C9H12ClNO2 | |
5 g | |
NDYXFQODWGEGNU-UHFFFAOYSA-N | |
2-(1,3-benzodioxol-5-yl)ethanamine;hydrochloride | |
74246 | |
95% |
Chemical Identifiers
1653-64-1 | |
201.65 | |
NDYXFQODWGEGNU-UHFFFAOYSA-N | |
74246 | |
C1OC2=C(O1)C=C(C=C2)CCN.Cl |
C9H12ClNO2 | |
MFCD00035193 | |
3,4-methylenedioxyphenethylamine hydrochloride, 2-3,4-methylenedioxyphenyl ethylamine hydrochloride, phenethylamine, 3,4-methylenedioxy-, hydrochloride, 2-benzo d 1,3 dioxol-5-yl ethanamine hydrochloride, homopiperonylamine hydrochloride, 1,3-benzodioxole-5-ethanamine, hydrochloride, 3,4-methylenedioxy-beta-phenylethylamine hydrochloride, methylenedioxyphenethylamine hydrochloride, 3,4-methylenedioxy phenylethylamine hydrochloride, 3,4-methylenedioxy-.beta.-phenylethylamine hydrochloride | |
2-(1,3-benzodioxol-5-yl)ethanamine;hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : SH9710000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only