missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(3,4-Epoxycyclohexyl)ethyltrimethoxysilane, TCI America™

Manufacturer:  TCI America E0327500ML

 View more versions of this product

Catalog No. E0327500ML



Specifications

Specifications

2-(3,4-Epoxycyclohexyl)ethyltrimethoxysilane
Colorless
3388-04-3
MFCD00014485
DQZNLOXENNXVAD-UHFFFAOYNA-N
trimethoxy-[2-(7-oxabicyclo[4.1.0]heptan-4-yl)ethyl]silane
18819
Clear Liquid at 20°C
310°C
500mL
C11H22O4Si
Trimethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane
CO[Si](CCC1CCC2C(C1)O2)(OC)OC
246.38
246.38
≥97.0% (GC)

Chemical Identifiers

3388-04-3
246.38
DQZNLOXENNXVAD-UHFFFAOYNA-N
18819
CO[Si](CCC1CCC2C(C1)O2)(OC)OC
C11H22O4Si
MFCD00014485
Trimethoxy[2-(7-oxabicyclo[4.1.0]heptan-3-yl)ethyl]silane
trimethoxy-[2-(7-oxabicyclo[4.1.0]heptan-4-yl)ethyl]silane
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2647

RTECSNumber : VV4000000

TSCA : Yes

Documents