missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3,4-Dimethoxyphenyl)ethyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ L1181703

 View more versions of this product

Catalog No. AAL1181703



Specifications

Specifications

2-(3,4-Dimethoxyphenyl)ethyl isothiocyanate
150°C to 151°C (0.3mmHg)
21714-25-0
MFCD00041366
2809316
GHYFLDWHIVKQLS-UHFFFAOYSA-N
4-(2-isothiocyanatoethyl)-1,2-dimethoxybenzene
140856
98%
Irritating
1g
C11H13NO2S
UN2922
3,4-dimethoxyphenethyl isothiocyanate, 4-2-isothiocyanatoethyl-1,2-dimethoxybenzene, 2-3,4-dimethoxyphenyl ethyl isothiocyanate, benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy, 2-3,4-dimethoxyphenyl ethanisothiocyanate, acmc-20anep, 3,4-dimethoxyphenethylisothiocyanate, b-3,4-dimethoxyphenyl ethyl isothiocyanate, 3,4-dimethoxy-1-2-isothiocyanatoethyl benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy-benzene
COC1=C(C=C(C=C1)CCN=C=S)OC
223.29
223.3

Chemical Identifiers

21714-25-0
223.29
GHYFLDWHIVKQLS-UHFFFAOYSA-N
140856
COC1=C(C=C(C=C1)CCN=C=S)OC
C11H13NO2S
MFCD00041366
3,4-dimethoxyphenethyl isothiocyanate, 4-2-isothiocyanatoethyl-1,2-dimethoxybenzene, 2-3,4-dimethoxyphenyl ethyl isothiocyanate, benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy, 2-3,4-dimethoxyphenyl ethanisothiocyanate, acmc-20anep, 3,4-dimethoxyphenethylisothiocyanate, b-3,4-dimethoxyphenyl ethyl isothiocyanate, 3,4-dimethoxy-1-2-isothiocyanatoethyl benzene, 4-2-isothiocyanatoethyl-1,2-dimethoxy-benzene
4-(2-isothiocyanatoethyl)-1,2-dimethoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.

Documents