Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3,4-Dimethoxyphenyl)acetamidoxime, 97%

Chemical Identifiers

Chemical Name or Material 2-(3,4-Dimethoxyphenyl)acetamidoxime
Formula Weight 210.23
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6681603

 
alfa aesar™
H6681603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6681606

 
alfa aesar™
H6681606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 210.23
Chemical Name or Material 2-(3,4-Dimethoxyphenyl)acetamidoxime
Physical Form Crystalline Solid
Assay 97%
Color White
Quantity 1g