Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,6-Tetrafluorobenzylamine hydrochloride, 98%

$161.62 - $624.95

Chemical Identifiers

Chemical Name or Material 2,3,4,6-Tetrafluorobenzylamine hydrochloride
Molecular Formula C7H5F4N·HCl
Formula Weight 215.58
MDL Number MFCD06199158
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2020703

  SDS
alfa aesar™
L2020703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2020706

  SDS
alfa aesar™
L2020706
5g Each for $161.62

AAL2020714

  SDS
alfa aesar™
L2020714
25g Each for $624.95
Description & Specifications

Specifications

Formula Weight 215.58
Chemical Name or Material 2,3,4,6-Tetrafluorobenzylamine hydrochloride
Assay 98%
Molecular Formula C7H5F4N·HCl
Health Hazard 1 Warning
MDL Number MFCD06199158
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 231° to 233°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.