Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,6-Tetrafluorobenzoyl chloride, 98%

Chemical Identifiers

Chemical Name or Material 2,3,4,6-Tetrafluorobenzoyl chloride
CAS 123016-51-3
Molecular Formula C7HClF4O
Formula Weight 212.53
MDL Number MFCD03001171
UN Number UN3265
Beilstein 6398940
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL2020606

 
alfa aesar™
L2020606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL2020614

 
alfa aesar™
L2020614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 212.53
Chemical Name or Material 2,3,4,6-Tetrafluorobenzoyl chloride
Assay 98%
Molecular Formula C7HClF4O
MDL Number MFCD03001171
Beilstein 6398940
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
CAS 123016-51-3
UN Number UN3265
Boiling Point 94° to 97°C (61mmHg)
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.