missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,6-Tetrafluorobenzeneboronic acid, 98%

$44.88 - $129.26

Chemical Identifiers

CAS 511295-00-4
Molecular Formula C6H3BF4O2
Formula Weight 193.9
MDL Number MFCD04039309
Beilstein 9200162
Synonym 2,3,4,6-Tetrafluorophenylboronic acid
Chemical Name or Material 2,3,4,6-Tetrafluorobenzeneboronic acid
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL19825MD SDS Alfa Aesar™
L19825MD
250mg Each for $44.88
AAL1982503 SDS Alfa Aesar™
L1982503
1g Each for $129.26
Specifications Description & Specifications

Chemical Identifiers

511295-00-4
193.9
9200162
2,3,4,6-Tetrafluorobenzeneboronic acid
C6H3BF4O2
MFCD04039309
2,3,4,6-Tetrafluorophenylboronic acid

Specifications

511295-00-4
193.9
9200162
≈125° to 129°C
2,3,4,6-Tetrafluorobenzeneboronic acid
C6H3BF4O2
MFCD04039309
2,3,4,6-Tetrafluorophenylboronic acid
250mg
98%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning