missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,6-Tetra-O-benzyl-D-galactopyranose, 98%

Manufacturer:  Alfa Aesar™ 4553106

 View more versions of this product

Catalog No. AA4553106



Specifications

Specifications

2,3,4,6-Tetra-O-benzyl-D-galactopyranose
5g
C34H36O6
540.65
98%
White
53081-25-7
MFCD00132927
Crystalline

Chemical Identifiers

53081-25-7
C34H36O6
MFCD00132927
Safety and Handling

Safety and Handling

Recommended Storage : −30°C to −10°C

SDS Documents