missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,4,6-Tetra-O-benzoyl-beta-D-glucopyranosyl Isothiocyanate [for HPLC Labeling], TCI America™

$192.50 - $1,147.39

Chemical Identifiers

CAS 132413-50-4
Molecular Formula C35H27NO9S
Molecular Weight (g/mol) 637.659
MDL Number MFCD00075064
InChI Key RITGYQZMPNVVEG-PXPWAULYSA-N
Synonym BGIT
PubChem CID 11966661
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-tribenzoyloxy-6-isothiocyanatooxan-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(C(O2)N=C=S)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4)OC(=O)C5=CC=CC=C5
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A5515100MG View Documents TCI America
A5515100MG
100mg Each for $192.50
 
A55151G View Documents TCI America
A55151G
1g Each for $1,147.39
 
Specifications

Chemical Identifiers

132413-50-4
637.659
RITGYQZMPNVVEG-PXPWAULYSA-N
11966661
C1=CC=C(C=C1)C(=O)OCC2C(C(C(C(O2)N=C=S)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4)OC(=O)C5=CC=CC=C5
C35H27NO9S
MFCD00075064
BGIT
[(2R,3R,4S,5R,6R)-3,4,5-tribenzoyloxy-6-isothiocyanatooxan-2-yl]methyl benzoate

Specifications

White-Yellow
132413-50-4
MFCD00075064
RITGYQZMPNVVEG-PXPWAULYSA-N
[(2R,3R,4S,5R,6R)-3,4,5-tribenzoyloxy-6-isothiocyanatooxan-2-yl]methyl benzoate
11966661
Crystal-Powder at 20°C
2,3,4,6-Tetra-O-benzoyl-beta-D-glucopyranosyl Isothiocyanate [for HPLC Labeling]
100mg
C35H27NO9S
BGIT
C1=CC=C(C=C1)C(=O)OCC2C(C(C(C(O2)N=C=S)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4)OC(=O)C5=CC=CC=C5
637.659
637.66
≥98.0% (HPLC)
Safety and Handling

Safety and Handling

TSCA : No