missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl Isothiocyanate [for HPLC Labeling], TCI America™

$151.25 - $818.91

Chemical Identifiers

CAS 14152-97-7
Molecular Formula C15H19NO9S
Molecular Weight (g/mol) 389.375
MDL Number MFCD00043085
InChI Key WOWNQYXIQWQJRJ-UXXRCYHCSA-N
Synonym GITC
PubChem CID 99462
IUPAC Name [(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-isothiocyanatooxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)N=C=S)OC(=O)C)OC(=O)C)OC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A5514100MG View Documents TCI America
A5514100MG
100mg Each for $151.25
 
A55141G View Documents TCI America
A55141G
1g Each for $818.91
 
Specifications

Chemical Identifiers

14152-97-7
389.375
WOWNQYXIQWQJRJ-UXXRCYHCSA-N
99462
CC(=O)OCC1C(C(C(C(O1)N=C=S)OC(=O)C)OC(=O)C)OC(=O)C
C15H19NO9S
MFCD00043085
GITC
[(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-isothiocyanatooxan-2-yl]methyl acetate

Specifications

White
100mg
C15H19NO9S
GITC
CC(=O)OCC1C(C(C(C(O1)N=C=S)OC(=O)C)OC(=O)C)OC(=O)C
389.375
389.38
≥98.0% (HPLC,N)
116°C
14152-97-7
MFCD00043085
WOWNQYXIQWQJRJ-UXXRCYHCSA-N
[(2R,3R,4S,5R,6R)-3,4,5-triacetyloxy-6-isothiocyanatooxan-2-yl]methyl acetate
99462
Crystal-Powder at 20°C
2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl Isothiocyanate [for HPLC Labeling]
Safety and Handling

Safety and Handling

TSCA : No