missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,6-Tetra-O-acetyl-alpha-D-mannopyranosyl fluoride, 98%

$112.72 - $504.27

Chemical Identifiers

CAS 2823-44-1
Molecular Formula C14H19FO9
Molecular Weight (g/mol) 350.295
MDL Number MFCD00792706
InChI Key JJXATNWYELAACC-DGTMBMJNSA-N
Synonym acetofluoro-alpha-d-mannose, 2,3,4,6-tetra-o-acetyl-alpha-d-mannopyranosyl fluoride, 2r,3r,4s,5s,6r-3,4,5-tris acetyloxy-6-fluorooxan-2-yl methyl acetate, acetofluoro-, a-d-mannose, tetra-o-acetyl-alpha-d-mannopyranosyl fluoride, 2,3,4,6-tetra-o-acetyl-, a-d-mannopyranosyl fluoride, 1-fluoro-1-deoxy-alpha-d-mannopyranose-2,3,4,6-tetraacetate, 2r,3r,4s,5s,6r-2-acetoxymethyl-6-fluorotetrahydro-2h-pyran-3,4,5-triyl triacetate
PubChem CID 11268124
IUPAC Name [(2R,3R,4S,5S,6R)-3,4,5-triacetyloxy-6-fluorooxan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(C(O1)F)OC(=O)C)OC(=O)C)OC(=O)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6613703 View Documents Alfa Aesar™
J6613703
1g Each for $112.72
 
AAJ6613706 View Documents Alfa Aesar™
J6613706
5g Each for $504.27
 
Specifications

Chemical Identifiers

2823-44-1
350.295
JJXATNWYELAACC-DGTMBMJNSA-N
11268124
CC(=O)OCC1C(C(C(C(O1)F)OC(=O)C)OC(=O)C)OC(=O)C
C14H19FO9
MFCD00792706
acetofluoro-alpha-d-mannose, 2,3,4,6-tetra-o-acetyl-alpha-d-mannopyranosyl fluoride, 2r,3r,4s,5s,6r-3,4,5-tris acetyloxy-6-fluorooxan-2-yl methyl acetate, acetofluoro-, a-d-mannose, tetra-o-acetyl-alpha-d-mannopyranosyl fluoride, 2,3,4,6-tetra-o-acetyl-, a-d-mannopyranosyl fluoride, 1-fluoro-1-deoxy-alpha-d-mannopyranose-2,3,4,6-tetraacetate, 2r,3r,4s,5s,6r-2-acetoxymethyl-6-fluorotetrahydro-2h-pyran-3,4,5-triyl triacetate
[(2R,3R,4S,5S,6R)-3,4,5-triacetyloxy-6-fluorooxan-2-yl]methyl acetate

Specifications

+21° (c=2 in Chloroform)
1g
C14H19FO9
acetofluoro-alpha-d-mannose, 2,3,4,6-tetra-o-acetyl-alpha-d-mannopyranosyl fluoride, 2r,3r,4s,5s,6r-3,4,5-tris acetyloxy-6-fluorooxan-2-yl methyl acetate, acetofluoro-, a-d-mannose, tetra-o-acetyl-alpha-d-mannopyranosyl fluoride, 2,3,4,6-tetra-o-acetyl-, a-d-mannopyranosyl fluoride, 1-fluoro-1-deoxy-alpha-d-mannopyranose-2,3,4,6-tetraacetate, 2r,3r,4s,5s,6r-2-acetoxymethyl-6-fluorotetrahydro-2h-pyran-3,4,5-triyl triacetate
CC(=O)OCC1C(C(C(C(O1)F)OC(=O)C)OC(=O)C)OC(=O)C
350.295
350.29
98%
White
2823-44-1
MFCD00792706
JJXATNWYELAACC-DGTMBMJNSA-N
[(2R,3R,4S,5S,6R)-3,4,5-triacetyloxy-6-fluorooxan-2-yl]methyl acetate
11268124
Powder
2,3,4,6-Tetra-O-acetyl-alpha-D-mannopyranosyl fluoride
Safety and Handling

Safety and Handling

Recommended Storage : −30° to −10°C