Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(3,4,5-Trifluorophenyl)ethanol, 96%

$38.73 - $132.28

Chemical Identifiers

Chemical Name or Material 2-(3,4,5-Trifluorophenyl)ethanol
CAS 886761-77-9
Molecular Formula C8H7F3O
Formula Weight 176.14
MDL Number MFCD06660355
Synonym 3,4,5-Trifluorophenethyl alcohol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3280003

  SDS
alfa aesar™
H3280003
1g Each for $132.28

AAH32800MD

  SDS
alfa aesar™
H32800MD
250mg Each for $38.73
Description & Specifications

Specifications

Formula Weight 176.14
CAS 886761-77-9
Quantity 1g
MDL Number MFCD06660355
Molecular Formula C8H7F3O
Chemical Name or Material 2-(3,4,5-Trifluorophenyl)ethanol
Assay 96%
Synonym 3,4,5-Trifluorophenethyl alcohol