missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3,4,5-Tetramethoxytoluene 97.0+%, TCI America™
Supplier: TCI America T262625G
Specifications
2,3,4,5-Tetramethoxytoluene | |
Yellow | |
C11H16O4 | |
25 g | |
CC1=CC(=C(C(=C1OC)OC)OC)OC | |
212.245 | |
212.25 | |
Liquid |
35896-58-3 | |
120°C | |
MFCD07370118 | |
OIWAVVSMXFIBCD-UHFFFAOYSA-N | |
1,2,3,4-tetramethoxy-5-methylbenzene | |
9877764 | |
≥97.0% (GC) |
Chemical Identifiers
35896-58-3 | |
212.245 | |
OIWAVVSMXFIBCD-UHFFFAOYSA-N | |
1,2,3,4-tetramethoxy-5-methylbenzene |
C11H16O4 | |
MFCD07370118 | |
9877764 | |
CC1=CC(=C(C(=C1OC)OC)OC)OC |
Safety and Handling
TSCA : No