missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,4,5-Tetrafluorobenzyl Bromide, TCI America™

Manufacturer:  TCI America T18855G

 View more versions of this product

Catalog No. T18855G



Specifications

Specifications

2,3,4,5-Tetrafluorobenzyl Bromide
Yellow
53001-71-1
MFCD01631453
2,3,4,5-tetrafluorobenzyl bromide, 1-bromomethyl-2,3,4,5-tetrafluorobenzene, 2,3,4,5-tetrafluorobenzylbromide, acmc-209l3c, 2,3,4,5-tetrafluorobenzyl bromide, alpha-bromo-2,3,4,5-tetrafluorotoluene, 1-bromomethyl-2,3,4,5-tetrafluoro-benzene, 5-bromomethyl-1,2,3,4-tetrafluorobenzene, benzene, 1-bromomethyl-2,3,4,5-tetrafluoro, 1-bromomethyl-2,3,4,5-tetrakis fluoranyl benzene
C1=C(C(=C(C(=C1F)F)F)F)CBr
242.999
243.00
≥97.0% (GC,T)
110°C
5g
C7H3BrF4
3265
WTKVWOHBTGWNEO-UHFFFAOYSA-N
1-(bromomethyl)-2,3,4,5-tetrafluorobenzene
2734024
Clear Liquid at 20°C

Chemical Identifiers

53001-71-1
242.999
WTKVWOHBTGWNEO-UHFFFAOYSA-N
2734024
C1=C(C(=C(C(=C1F)F)F)F)CBr
C7H3BrF4
MFCD01631453
2,3,4,5-tetrafluorobenzyl bromide, 1-bromomethyl-2,3,4,5-tetrafluorobenzene, 2,3,4,5-tetrafluorobenzylbromide, acmc-209l3c, 2,3,4,5-tetrafluorobenzyl bromide, alpha-bromo-2,3,4,5-tetrafluorotoluene, 1-bromomethyl-2,3,4,5-tetrafluoro-benzene, 5-bromomethyl-1,2,3,4-tetrafluorobenzene, benzene, 1-bromomethyl-2,3,4,5-tetrafluoro, 1-bromomethyl-2,3,4,5-tetrakis fluoranyl benzene
1-(bromomethyl)-2,3,4,5-tetrafluorobenzene
Safety and Handling

Safety and Handling

TSCA : No

Documents