Please login to your online account to display your discounted pricing

2,3,4,5-Tetrafluorobenzyl bromide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2,3,4,5-Tetrafluorobenzyl bromide
Name Note 98%
CAS 53001-71-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H3BrF4
Formula Weight 242.99
MDL Number MFCD01631453
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC347430010

  SDS
thermo fisher scientific
347430010
Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5%
Formula Weight 242.99
Quantity 1g
Color Clear colorless to yellow
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 2,3,4,5-Tetrafluorobenzyl bromide
Infrared Spectrum Authentic
Refractive Index 1.4890 to 1.4910 (20°C, 589nm)
Name Note 98%
Assay Percent Range 98%
Molecular Formula C7H3BrF4
CAS 53001-71-1
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Packaging Glass Bottle
MDL Number MFCD01631453
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.