missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2,3,4,5-Tetrafluorobenzyl bromide, 97%

$81.58 - $255.99

Chemical Identifiers

CAS 53001-71-1
Molecular Formula C7H3BrF4
Molecular Weight (g/mol) 242.999
MDL Number MFCD01631453
InChI Key WTKVWOHBTGWNEO-UHFFFAOYSA-N
Synonym 2,3,4,5-tetrafluorobenzyl bromide, 1-bromomethyl-2,3,4,5-tetrafluorobenzene, 2,3,4,5-tetrafluorobenzylbromide, acmc-209l3c, 2,3,4,5-tetrafluorobenzyl bromide, alpha-bromo-2,3,4,5-tetrafluorotoluene, 1-bromomethyl-2,3,4,5-tetrafluoro-benzene, 5-bromomethyl-1,2,3,4-tetrafluorobenzene, benzene, 1-bromomethyl-2,3,4,5-tetrafluoro, 1-bromomethyl-2,3,4,5-tetrakis fluoranyl benzene
PubChem CID 2734024
IUPAC Name 1-(bromomethyl)-2,3,4,5-tetrafluorobenzene
SMILES C1=C(C(=C(C(=C1F)F)F)F)CBr
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1816203 View Documents Alfa Aesar™
L1816203
1g Each for $81.58
 
AAL1816206 View Documents Alfa Aesar™
L1816206
5g Each for $255.99
 
Specifications

Chemical Identifiers

53001-71-1
242.999
WTKVWOHBTGWNEO-UHFFFAOYSA-N
2734024
C1=C(C(=C(C(=C1F)F)F)F)CBr
C7H3BrF4
MFCD01631453
2,3,4,5-tetrafluorobenzyl bromide, 1-bromomethyl-2,3,4,5-tetrafluorobenzene, 2,3,4,5-tetrafluorobenzylbromide, acmc-209l3c, 2,3,4,5-tetrafluorobenzyl bromide, alpha-bromo-2,3,4,5-tetrafluorotoluene, 1-bromomethyl-2,3,4,5-tetrafluoro-benzene, 5-bromomethyl-1,2,3,4-tetrafluorobenzene, benzene, 1-bromomethyl-2,3,4,5-tetrafluoro, 1-bromomethyl-2,3,4,5-tetrakis fluoranyl benzene
1-(bromomethyl)-2,3,4,5-tetrafluorobenzene

Specifications

107°C to 110°C (65mmHg)
53001-71-1
MFCD01631453
2263035
WTKVWOHBTGWNEO-UHFFFAOYSA-N
1-(bromomethyl)-2,3,4,5-tetrafluorobenzene
2734024
97%
1g
C7H3BrF4
UN3265
2,3,4,5-tetrafluorobenzyl bromide, 1-bromomethyl-2,3,4,5-tetrafluorobenzene, 2,3,4,5-tetrafluorobenzylbromide, acmc-209l3c, 2,3,4,5-tetrafluorobenzyl bromide, alpha-bromo-2,3,4,5-tetrafluorotoluene, 1-bromomethyl-2,3,4,5-tetrafluoro-benzene, 5-bromomethyl-1,2,3,4-tetrafluorobenzene, benzene, 1-bromomethyl-2,3,4,5-tetrafluoro, 1-bromomethyl-2,3,4,5-tetrakis fluoranyl benzene
C1=C(C(=C(C(=C1F)F)F)F)CBr
242.999
243
2,3,4,5-Tetrafluorobenzyl bromide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.