missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,3,4,5-Tetrafluorobenzoyl Chloride 97.0+%, TCI America™
Supplier: TCI America T185525G
Specifications
2,3,4,5-Tetrafluorobenzoyl Chloride | |
Yellow | |
MFCD00075164 | |
3265 | |
XWCKIXLTBNGIHV-UHFFFAOYSA-N | |
2,3,4,5-tetrafluorobenzoyl chloride | |
2733689 | |
≥97.0% (GC,T) |
94695-48-4 | |
C7HClF4O | |
25 g | |
2,3,4,5-tetrafluorobenzoylchloride, tetrafluorobenzoyl chloride, 2,3,4,5-tetrafluoro benzoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro, 2,3,4,5-tetrafluorbenzoylchlorid, 2,3,4,5-tetrafluoro-benzoyl chloride, 2,3,4,5-tetrafluorobenzoic acid chloride, 2,3,4,5-tetrafluorobezoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro-9ci, 2,3,4,5-tetrafluorobezoylchloride | |
FC1=CC(C(Cl)=O)=C(F)C(F)=C1F | |
212.53 | |
212.53 | |
Liquid |
Chemical Identifiers
94695-48-4 | |
212.53 | |
XWCKIXLTBNGIHV-UHFFFAOYSA-N | |
2733689 | |
FC1=CC(C(Cl)=O)=C(F)C(F)=C1F |
C7HClF4O | |
MFCD00075164 | |
2,3,4,5-tetrafluorobenzoylchloride, tetrafluorobenzoyl chloride, 2,3,4,5-tetrafluoro benzoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro, 2,3,4,5-tetrafluorbenzoylchlorid, 2,3,4,5-tetrafluoro-benzoyl chloride, 2,3,4,5-tetrafluorobenzoic acid chloride, 2,3,4,5-tetrafluorobezoyl chloride, benzoyl chloride, 2,3,4,5-tetrafluoro-9ci, 2,3,4,5-tetrafluorobezoylchloride | |
2,3,4,5-tetrafluorobenzoyl chloride |
Safety and Handling
TSCA : No