missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,3,4,5-Tetrafluorobenzoyl chloride, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 334570010

 View more versions of this product

Catalog No. AC334570010

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2,3,4,5-Tetrafluorobenzoyl chloride
Clear colorless
99%
98.5
Authentic
1.4780 to 1.4800 (20°C, 589nm)
≥98.5%
C7 H Cl F4 O
MFCD00075164
1.5800g/mL
1g
94695-48-4
100.0
Glass bottle
Liquid
99%
212.53

Chemical Identifiers

94695-48-4
100.0
C7 H Cl F4 O
MFCD00075164
≥98.5%
98.5
99%
212.53
2,3,4,5-Tetrafluorobenzoyl chloride, 99%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA